siRNA Details
| Gene Symbol: Abcb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080582.1 | NP_542149.1 | 4 | 1085 | ||
| GenBank mRNA | AJ003004.1 | - | 4 | 1085 | ||
| BC085712.1 | - | 4 | 1101 | 7190755 | ||
| FQ211570.1 | - | 4 | 1111 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcb6| Gene Symbol: Abcb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080582.1 | NP_542149.1 | 4 | 1085 | ||
| GenBank mRNA | AJ003004.1 | - | 4 | 1085 | ||
| BC085712.1 | - | 4 | 1101 | 7190755 | ||
| FQ211570.1 | - | 4 | 1111 | |||