siRNA Details
| Gene Symbol: Dnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080689.4 | NP_542420.1 | 8 | 1080 | ||
| GenBank mRNA | X54531.1 | - | 8 | 1080 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnm1| Gene Symbol: Dnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080689.4 | NP_542420.1 | 8 | 1080 | ||
| GenBank mRNA | X54531.1 | - | 8 | 1080 | ||