siRNA Details
| Gene Symbol: Mmp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080776.1 | NP_542954.1 | 5 | 1270 | ||
| XM_006237921.2 | XP_006237983.1 | 5 | 1336 | |||
| GenBank mRNA | D63886.1 | - | 5 | 1270 | ||
| D85509.1 | - | 5 | 1270 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmp16| Gene Symbol: Mmp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080776.1 | NP_542954.1 | 5 | 1270 | ||
| XM_006237921.2 | XP_006237983.1 | 5 | 1336 | |||
| GenBank mRNA | D63886.1 | - | 5 | 1270 | ||
| D85509.1 | - | 5 | 1270 | |||