siRNA Details
| Gene Symbol: Msmo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080886.1 | NP_543162.1 | 4 | 577 | ||
| GenBank mRNA | BC063155.1 | - | 4 | 564 | 6918081 | |
| D50559.1 | - | 4 | 577 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msmo1| Gene Symbol: Msmo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080886.1 | NP_543162.1 | 4 | 577 | ||
| GenBank mRNA | BC063155.1 | - | 4 | 564 | 6918081 | |
| D50559.1 | - | 4 | 577 | |||