siRNA Details
| Gene Symbol: Sdha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130428.1 | NP_569112.1 | 15 | 2044 | ||
| GenBank mRNA | AB072907.1 | - | 15 | 2044 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sdha| Gene Symbol: Sdha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130428.1 | NP_569112.1 | 15 | 2044 | ||
| GenBank mRNA | AB072907.1 | - | 15 | 2044 | ||