siRNA Details
| Gene Symbol: Gucy2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130737.4 | NP_570093.1 | 4 | 1272 | ||
| XM_006229710.1 | XP_006229772.1 | 4 | 1271 | |||
| GenBank mRNA | L37203.1 | - | 4 | 1272 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gucy2d| Gene Symbol: Gucy2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130737.4 | NP_570093.1 | 4 | 1272 | ||
| XM_006229710.1 | XP_006229772.1 | 4 | 1271 | |||
| GenBank mRNA | L37203.1 | - | 4 | 1272 | ||