siRNA Details
| Gene Symbol: Acsl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130739.1 | NP_570095.1 | 21 | 2419 | ||
| GenBank mRNA | S56508.1 | - | 21 | 2419 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acsl6| Gene Symbol: Acsl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130739.1 | NP_570095.1 | 21 | 2419 | ||
| GenBank mRNA | S56508.1 | - | 21 | 2419 | ||