siRNA Details
Gene Symbol: Cygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130744.2 | NP_570100.1 | 4 | 1148 | ||
GenBank mRNA | AJ245663.2 | - | 4 | 1056 | ||
BC088455.1 | - | 4 | 1148 | 7127197 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130744.2 | NP_570100.1 | 4 | 1148 | ||
GenBank mRNA | AJ245663.2 | - | 4 | 1056 | ||
BC088455.1 | - | 4 | 1148 | 7127197 |