siRNA Details
| Gene Symbol: Slc5a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130746.1 | NP_570102.1 | 4 | 606 | ||
| GenBank mRNA | AF026554.1 | - | 4 | 606 | ||
| AF081204.2 | - | 1 | 194 | |||
| AF081205.1 | - | 1 | 194 | |||
| BC168145.1 | - | 2 | 468 | 8371761 | ||

Slc5a6
Genomic Map