siRNA Details
| Gene Symbol: Slc5a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130746.1 | NP_570102.1 | 10, 11 | 1419 | ||
| GenBank mRNA | AF026554.1 | - | 10, 11 | 1419 | ||
| AF081204.2 | - | 7, 8 | 1007 | |||
| AF081205.1 | - | 7, 8 | 1007 | |||
| BC168145.1 | - | 8, 9 | 1281 | 8371761 | ||

Slc5a6
Genomic Map