siRNA Details
Gene Symbol: Pnma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130820.2 | NP_570833.1 | 1 | 600 | ||
GenBank mRNA | AF335505.1 | - | 1 | 616 | ||
BC097291.1 | - | 1 | 576 | 7453131 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pnma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130820.2 | NP_570833.1 | 1 | 600 | ||
GenBank mRNA | AF335505.1 | - | 1 | 616 | ||
BC097291.1 | - | 1 | 576 | 7453131 |