siRNA Details
Gene Symbol: Atp2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_131907.2 | NP_571982.2 | 4 | 683 | ||
GenBank mRNA | BC086994.1 | - | 4 | 683 | 7106233 | |
M93017.1 | - | 4 | 394 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atp2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_131907.2 | NP_571982.2 | 4 | 683 | ||
GenBank mRNA | BC086994.1 | - | 4 | 683 | 7106233 | |
M93017.1 | - | 4 | 394 |