siRNA Details
| Gene Symbol: Bhlhe41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729454.5 | XP_002729500.2 | 4 | 639 | ||
| GenBank mRNA | AF009329.1 | - | 4 | 537 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bhlhe41| Gene Symbol: Bhlhe41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729454.5 | XP_002729500.2 | 4 | 639 | ||
| GenBank mRNA | AF009329.1 | - | 4 | 537 | ||