siRNA Details
| Gene Symbol: Heph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133304.1 | NP_579838.1 | 3 | 349 | ||
| XM_006257046.2 | XP_006257108.1 | 4 | 649 | |||
| XM_006257048.2 | XP_006257110.1 | 4 | 884 | |||
| XM_006257049.2 | XP_006257111.1 | 3 | 684 | |||
| XM_008773248.1 | XP_008771470.1 | 5 | 1282 | |||
| XM_008773249.1 | XP_008771471.1 | 5 | 978 | |||
| XM_008773250.1 | XP_008771472.1 | 4 | 652 | |||
| GenBank mRNA | AF246120.1 | - | 3 | 349 | ||

Heph
Genomic Map