siRNA Details
| Gene Symbol: Tmlhe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133387.2 | NP_596878.2 | 4 | 438 | ||
| XM_006255855.1 | XP_006255917.1 | 3 | 447 | |||
| GenBank mRNA | AF374406.1 | - | 2 | 279 | ||
| BC085923.1 | - | 4 | 438 | 7109209 | ||

Tmlhe
Genomic Map