siRNA Details
| Gene Symbol: Kcng3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033957.1 | NP_001029129.1 | 2 | 684 | ||
| NM_133426.2 | NP_596917.2 | 2 | 717 | |||
| GenBank mRNA | AB070605.1 | - | 2 | 411 | ||
| AF454549.1 | - | 2 | 868 | |||
| AF454550.1 | - | 2 | 901 | |||

Kcng3
Genomic Map