siRNA Details
| Gene Symbol: Hrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133428.2 | NP_596919.1 | 1 | 124 | ||
| GenBank mRNA | AB055895.1 | - | 1 | 124 | ||
| AB055896.1 | - | 1 | 115 | |||
| AF194029.1 | - | 1 | 79 | |||
| BC089779.1 | - | 1 | 113 | 7371893 | ||
| FQ210661.1 | - | 1 | 124 | |||

Hrg
Genomic Map