siRNA Details
| Gene Symbol: Fbxo32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133521.1 | NP_598205.1 | 6 | 470 | ||
| GenBank mRNA | AY059628.1 | - | 6 | 470 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo32| Gene Symbol: Fbxo32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133521.1 | NP_598205.1 | 6 | 470 | ||
| GenBank mRNA | AY059628.1 | - | 6 | 470 | ||