siRNA Details
Gene Symbol: Folr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133527.1 | NP_598211.1 | 5 | 890 | ||
XM_006229842.2 | XP_006229904.1 | 5 | 1012 | |||
GenBank mRNA | AF219904.1 | - | 5 | 890 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Folr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133527.1 | NP_598211.1 | 5 | 890 | ||
XM_006229842.2 | XP_006229904.1 | 5 | 1012 | |||
GenBank mRNA | AF219904.1 | - | 5 | 890 |