siRNA Details
Gene Symbol: Gtf3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133541.1 | NP_598225.1 | 2 | 432 | ||
XM_006230205.2 | XP_006230267.1 | 2 | 453 | |||
GenBank mRNA | L28801.1 | - | 2 | 432 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gtf3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133541.1 | NP_598225.1 | 2 | 432 | ||
XM_006230205.2 | XP_006230267.1 | 2 | 453 | |||
GenBank mRNA | L28801.1 | - | 2 | 432 |