siRNA Details
| Gene Symbol: Gtf3c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133541.1 | NP_598225.1 | 11 | 1878 | ||
| XM_006230205.2 | XP_006230267.1 | 11 | 1899 | |||
| XM_008759857.1 | XP_008758079.1 | 5 | 849 | |||
| GenBank mRNA | L28801.1 | - | 11 | 1878 | ||

Gtf3c1
Genomic Map