siRNA Details
| Gene Symbol: FBXL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193534.1 | NP_001180463.1 | 2 | 289 | ||
| NM_001193535.1 | NP_001180464.1 | 2 | 238 | |||
| NM_012161.3 | NP_036293.1 | 2 | 289 | |||
| NR_036464.1 | 2 | 289 | ||||
| XM_006713959.3 | XP_006714022.1 | 3 | 385 | |||
| XM_011513831.2 | XP_011512133.1 | 2 | 71 | |||
| XM_011513832.2 | XP_011512134.1 | 2 | 218 | |||
| XM_017008019.1 | XP_016863508.1 | 4 | 1156 | |||
| GenBank mRNA | AF142481.1 | - | 2 | 211 | ||
| AF157323.1 | - | 2 | 150 | |||
| AF176700.1 | - | 2 | 173 | |||
| AF199420.1 | - | 2 | 178 | |||
| AK295510.1 | - | 2 | 208 | |||
| AK310860.1 | - | 2 | 218 | |||
| AL832808.1 | - | 1 | 21 | |||
| BC012320.1 | - | 2 | 218 | |||
| BI464829.1 | - | 2 | 268 | |||
| BX537957.1 | - | 2 | 184 | |||
| DB080171.1 | - | 2 | 290 | |||

FBXL5
Genomic Map