siRNA Details
Gene Symbol: Pde5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133584.1 | NP_598268.1 | 2 | 937 | ||
XM_006233260.2 | XP_006233322.1 | 2 | 865 | |||
GenBank mRNA | D89093.1 | - | 2 | 937 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pde5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133584.1 | NP_598268.1 | 2 | 937 | ||
XM_006233260.2 | XP_006233322.1 | 2 | 865 | |||
GenBank mRNA | D89093.1 | - | 2 | 937 |