siRNA Details
| Gene Symbol: Rhov | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138542.2 | NP_612551.2 | 4 | 593 | ||
| XM_008762077.1 | XP_008760299.1 | 4 | 900 | |||
| GenBank mRNA | AF097887.1 | - | 3 | 548 | ||
| BC086990.1 | - | 4 | 593 | 7135643 | ||

Rhov
Genomic Map