siRNA Details
| Gene Symbol: Snrk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138833.1 | NP_620188.1 | 2 | 351 | ||
| XM_006244057.2 | XP_006244119.1 | 3 | 397 | |||
| XM_006244058.2 | XP_006244120.1 | 3 | 431 | |||
| XM_006244059.1 | XP_006244121.1 | 3 | 390 | |||
| XM_006244060.2 | XP_006244122.1 | 2 | 405 | |||
| XM_006244061.1 | XP_006244123.1 | 2 | 411 | |||
| XM_006244062.2 | XP_006244124.1 | 2 | 339 | |||
| XM_008766666.1 | XP_008764888.1 | 2 | 330 | |||
| XM_008766667.1 | XP_008764889.1 | 2 | 472 | |||
| GenBank mRNA | X89383.1 | - | 2 | 351 | ||

Snrk
Genomic Map