siRNA Details
| Gene Symbol: Ick | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138886.1 | NP_620241.1 | 7 | 927 | ||
| XM_008766345.1 | XP_008764567.1 | 7 | 1150 | |||
| GenBank mRNA | D26178.1 | - | 7 | 927 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ick| Gene Symbol: Ick | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138886.1 | NP_620241.1 | 7 | 927 | ||
| XM_008766345.1 | XP_008764567.1 | 7 | 1150 | |||
| GenBank mRNA | D26178.1 | - | 7 | 927 | ||