siRNA Details
| Gene Symbol: Cdipt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138899.2 | NP_620254.1 | 6 | 1786 | ||
| XM_006230208.1 | XP_006230270.1 | 7 | 1822 | |||
| GenBank mRNA | BC070876.1 | - | 6 | 1786 | 7096385 | |
| D82928.1 | - | 6 | 1534 | |||

Cdipt
Genomic Map