siRNA Details
Gene Symbol: Mfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138976.1 | NP_620432.1 | 2 | 182 | ||
GenBank mRNA | AB084166.1 | - | 2 | 182 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138976.1 | NP_620432.1 | 2 | 182 | ||
GenBank mRNA | AB084166.1 | - | 2 | 182 |