siRNA Details
| Gene Symbol: Uxs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139336.1 | NP_647552.1 | 6 | 471 | ||
| XM_006244296.2 | XP_006244358.1 | 6 | 529 | |||
| XM_008766746.1 | XP_008764968.1 | 7 | 987 | |||
| XM_008766747.1 | XP_008764969.1 | 639 | ||||
| XM_008766748.1 | XP_008764970.1 | 7 | 972 | |||
| XM_008766749.1 | XP_008764971.1 | 6 | 531 | |||
| XM_008766750.1 | XP_008764972.1 | 6 | 516 | |||
| XM_008766751.1 | XP_008764973.1 | 7 | 987 | |||
| XM_008766752.1 | XP_008764974.1 | 6 | 879 | |||
| XM_008766753.1 | XP_008764975.1 | 6 | 864 | |||
| GenBank mRNA | AF482705.1 | - | 6 | 471 | ||
| BC086988.1 | - | 6 | 482 | 7192831 | ||

Uxs1
Genomic Map