siRNA Details
| Gene Symbol: Usp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145184.1 | NP_660185.1 | 1, 2 | 129 | ||
| XM_006241431.2 | XP_006241493.1 | 1, 2 | 160 | |||
| GenBank mRNA | AF106657.1 | - | 1, 2 | 129 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp15| Gene Symbol: Usp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145184.1 | NP_660185.1 | 1, 2 | 129 | ||
| XM_006241431.2 | XP_006241493.1 | 1, 2 | 160 | |||
| GenBank mRNA | AF106657.1 | - | 1, 2 | 129 | ||