siRNA Details
| Gene Symbol: Usp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145184.1 | NP_660185.1 | 9 | 1138 | ||
| XM_006241431.2 | XP_006241493.1 | 10 | 1256 | |||
| XM_008765376.1 | XP_008763598.1 | 8 | 1568 | |||
| GenBank mRNA | AF106657.1 | - | 9 | 1138 | ||

Usp15
Genomic Map