siRNA Details
| Gene Symbol: Acox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145770.2 | NP_665713.2 | 3 | 407 | ||
| XM_006251743.2 | XP_006251805.1 | 4 | 492 | |||
| XM_006251744.2 | XP_006251806.1 | 3 | 363 | |||
| XM_006251745.2 | XP_006251807.1 | 4 | 410 | |||
| GenBank mRNA | X95189.1 | - | 3 | 406 | ||

Acox2
Genomic Map