siRNA Details
Gene Symbol: Olfm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145777.1 | NP_665720.1 | 4 | 626 | ||
GenBank mRNA | AF442822.1 | - | 4 | 626 | ||
AF442823.1 | - | 517 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145777.1 | NP_665720.1 | 4 | 626 | ||
GenBank mRNA | AF442822.1 | - | 4 | 626 | ||
AF442823.1 | - | 517 |