siRNA Details
| Gene Symbol: Nr1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147210.2 | NP_671743.2 | 4 | 599 | ||
| GenBank mRNA | U15660.1 | - | 3 | 316 | ||
| U20796.1 | - | 3 | 256 | |||
| X78135.1 | - | 4 | 452 | |||
| X82777.1 | - | 4 | 534 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr1d2| Gene Symbol: Nr1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147210.2 | NP_671743.2 | 4 | 599 | ||
| GenBank mRNA | U15660.1 | - | 3 | 316 | ||
| U20796.1 | - | 3 | 256 | |||
| X78135.1 | - | 4 | 452 | |||
| X82777.1 | - | 4 | 534 | |||