siRNA Details
| Gene Symbol: Rcan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153724.2 | NP_714946.2 | 4 | 1238 | ||
| XM_006248041.2 | XP_006248103.1 | 4 | 1438 | |||
| GenBank mRNA | BC072478.1 | - | 4 | 1238 | 7103498 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rcan1| Gene Symbol: Rcan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153724.2 | NP_714946.2 | 4 | 1238 | ||
| XM_006248041.2 | XP_006248103.1 | 4 | 1438 | |||
| GenBank mRNA | BC072478.1 | - | 4 | 1238 | 7103498 | |