siRNA Details
Gene Symbol: Pex14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172063.1 | NP_742060.1 | 5 | 376 | ||
GenBank mRNA | AB017544.1 | - | 5 | 376 | ||
BC081708.1 | - | 5 | 365 | 7191447 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pex14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172063.1 | NP_742060.1 | 5 | 376 | ||
GenBank mRNA | AB017544.1 | - | 5 | 376 | ||
BC081708.1 | - | 5 | 365 | 7191447 |