siRNA Details
| Gene Symbol: Tfpi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173141.1 | NP_775164.1 | 3 | 452 | ||
| GenBank mRNA | AJ428954.1 | - | 3 | 452 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tfpi2| Gene Symbol: Tfpi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173141.1 | NP_775164.1 | 3 | 452 | ||
| GenBank mRNA | AJ428954.1 | - | 3 | 452 | ||