siRNA Details
| Gene Symbol: Wnk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175579.3 | NP_783169.2 | 1 | 403 | ||
| XM_006247278.2 | XP_006247340.1 | 1 | 508 | |||
| XM_006247279.2 | XP_006247341.1 | 1 | 508 | |||
| GenBank mRNA | AY187039.1 | - | 1 | 380 | ||
| AY192567.1 | - | 1 | 403 | |||

Wnk4
Genomic Map