siRNA Details
| Gene Symbol: Wnk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175579.3 | NP_783169.2 | 19 | 3969 | ||
| XM_006247278.2 | XP_006247340.1 | 18 | 4181 | |||
| XM_006247279.2 | XP_006247341.1 | 18 | 4178 | |||
| XM_006247280.2 | XP_006247342.1 | 19 | 3588 | |||
| XM_006247281.2 | XP_006247343.1 | 17 | 3340 | |||
| GenBank mRNA | AY187039.1 | - | 19 | 3946 | ||

Wnk4
Genomic Map