siRNA Details
| Gene Symbol: Id4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175582.1 | NP_783172.1 | 3 | 721 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Id4| Gene Symbol: Id4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175582.1 | NP_783172.1 | 3 | 721 | ||