siRNA Details
| Gene Symbol: Cryl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175757.2 | NP_786933.1 | 2 | 185 | ||
| GenBank mRNA | AY040223.1 | - | 2 | 185 | ||
| BC078685.1 | - | 2 | 159 | 7110482 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cryl1| Gene Symbol: Cryl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175757.2 | NP_786933.1 | 2 | 185 | ||
| GenBank mRNA | AY040223.1 | - | 2 | 185 | ||
| BC078685.1 | - | 2 | 159 | 7110482 | ||