siRNA Details
| Gene Symbol: Ugt1a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175846.2 | NP_787040.2 | 1 | 296 | ||
| GenBank mRNA | AF461735.1 | - | 1 | 299 | ||
| AY435135.1 | - | 1 | 296 | |||
| Gene Symbol: LOC102553187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767335.1 | XP_008765557.1 | 2 | 3135 | ||
| XM_008767336.1 | XP_008765558.1 | 2 | 3034 | |||

Ugt1a8
Genomic Map