siRNA Details
| Gene Symbol: Ppargc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176075.2 | NP_788264.1 | 175 | |||
| GenBank mRNA | AY188951.1 | - | 175 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppargc1b| Gene Symbol: Ppargc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176075.2 | NP_788264.1 | 175 | |||
| GenBank mRNA | AY188951.1 | - | 175 | |||