siRNA Details
| Gene Symbol: Kcnmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176861.1 | NP_789831.1 | 4 | 694 | ||
| GenBank mRNA | AB193522.1 | - | 2 | 228 | ||
| AB193523.1 | - | 2 | 327 | |||
| AY191836.1 | - | 4 | 694 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnmb2| Gene Symbol: Kcnmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176861.1 | NP_789831.1 | 4 | 694 | ||
| GenBank mRNA | AB193522.1 | - | 2 | 228 | ||
| AB193523.1 | - | 2 | 327 | |||
| AY191836.1 | - | 4 | 694 | |||