siRNA Details
| Gene Symbol: MID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012216.3 | NP_036348.2 | 2 | 820 | ||
| NM_052817.2 | NP_438112.2 | 2 | 820 | |||
| XM_005262062.4 | XP_005262119.1 | 2 | 5083 | |||
| XM_017029239.1 | XP_016884728.1 | 2 | 5083 | |||
| GenBank mRNA | AF196481.1 | - | 2 | 260 | ||
| AY625004.1 | - | 1 | 187 | |||
| BC017707.1 | - | 1 | 214 | 4446324 | ||
| BT006663.1 | - | 1 | 187 | |||
| Y18880.1 | - | 2 | 444 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011845, Y18881, AF196480.

MID2
Genomic Map