siRNA Details
| Gene Symbol: Vac14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177930.2 | NP_808791.1 | 3 | 592 | ||
| GenBank mRNA | AY220476.1 | - | 3 | 592 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vac14| Gene Symbol: Vac14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177930.2 | NP_808791.1 | 3 | 592 | ||
| GenBank mRNA | AY220476.1 | - | 3 | 592 | ||