siRNA Details
| Gene Symbol: Pafah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177932.2 | NP_808793.1 | 9 | 1039 | ||
| XM_006239127.2 | XP_006239189.1 | 10 | 1340 | |||
| XM_008764144.1 | XP_008762366.1 | 10 | 1342 | |||
| GenBank mRNA | AY225592.1 | - | 9 | 1039 | ||

Pafah2
Genomic Map