siRNA Details
| Gene Symbol: Abca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178095.2 | NP_835196.1 | 9 | 1179 | ||
| GenBank mRNA | AY208182.1 | - | 9 | 1179 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abca1| Gene Symbol: Abca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178095.2 | NP_835196.1 | 9 | 1179 | ||
| GenBank mRNA | AY208182.1 | - | 9 | 1179 | ||