siRNA Details
Gene Symbol: Plod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178101.3 | NP_835202.2 | 5 | 869 | ||
GenBank mRNA | AJ430859.1 | - | 5 | 608 | ||
BC085683.1 | - | 5 | 649 | 7110561 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Plod3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178101.3 | NP_835202.2 | 5 | 869 | ||
GenBank mRNA | AJ430859.1 | - | 5 | 608 | ||
BC085683.1 | - | 5 | 649 | 7110561 |