siRNA Details
| Gene Symbol: Mapkapk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178102.2 | NP_835203.1 | 2 | 528 | ||
| GenBank mRNA | AY197741.1 | - | 2 | 528 | ||
| BC062048.1 | - | 2 | 494 | 5598069 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mapkapk2| Gene Symbol: Mapkapk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178102.2 | NP_835203.1 | 2 | 528 | ||
| GenBank mRNA | AY197741.1 | - | 2 | 528 | ||
| BC062048.1 | - | 2 | 494 | 5598069 | ||